
N-Nitrosoethylisopropylamine (NEIPA)

Synonym(s):
N-Ethyl-N-nitroso-2-propanamine, EIPNA, NEIPA
Empirical Formula (Hill Notation):
C5H12N2O
CAS Number:
Molecular Weight:
116.16
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing