Skip to Content
Merck
N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride protease substrate

N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride

N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride protease substrate
Synonym(s):
N-Benzoyl-L-phenylalanyl-L-valyl-L-arginine-4-nitroanilide hydrochloride
Empirical Formula (Hill Notation):
C33H40N8O6 · HCl
CAS Number:
Molecular Weight:
681.18

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment