
Levocabastine for system suitability 2

Synonym(s):
Levocabastine hydrochloride, Levophta, Levostin, R50547, [3S-[1(cis),3α,4β]]-1-[4-cyano-4-(4-fluorophenyl)cyclohexyl]-3-methyl-4-phenyl-4-piperidinecarboxylic acid
Empirical Formula (Hill Notation):
C26H29N2O2F·HCl
CAS Number:
Molecular Weight:
456.98
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing