
Glutathione Impurity C

Synonym(s):
(−)-Glutathione, oxidized, L-Glutathione oxidized, GSSG, Glutathiol
Linear Formula:
[HO2CCH(NH2)CH2CH2CONHCH(CONHCH2CO2H)CH2S-]2
CAS Number:
Molecular Weight:
612.63
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing