
Glucotropaeolin potassium salt

Synonym(s):
Benzylglucosinolate potassium salt
Empirical Formula (Hill Notation):
C14H18KNO9S2
CAS Number:
Molecular Weight:
447.52
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing