Skip to Content
Merck
Ethylamine solution 2.0 M in methanol

Ethylamine solution

Ethylamine solution 2.0 M in methanol
Synonym(s):
Aminoethane, Monoethylamine
Linear Formula:
C2H5NH2
CAS Number:
Molecular Weight:
45.08

Products

Product Category
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing