
Elastatinal

Synonym(s):
(2S)-2-[(4S)-2-Amino-1,4,5,6-tetrahydro4-pyrimidinyl]-N-[[[(1S)-1-carboxy-3-methylbutyl]amino]carbonyl]glycyl-N1-[(1S)-1-methyl-2-oxoethyl-L-glutamamide
Empirical Formula (Hill Notation):
C21H36N8O7
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing