Skip to Content
Merck
Dihexadecyldimethylammonium bromide 97%

Dihexadecyldimethylammonium bromide

Dihexadecyldimethylammonium bromide 97%
Synonym(s):
Dimethyldihexadecylammonium bromide
Linear Formula:
[CH3(CH2)15]2N(CH3)2(Br)
CAS Number:
Molecular Weight:
574.85
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing