
Desflurane Related Compound A

Synonym(s):
1,1′-Oxybis(1,2,2,2-tetrafluoroethane), Bis(1,2,2,2-tetrafluoroethyl) ether
Empirical Formula (Hill Notation):
C4H2F8O
CAS Number:
Molecular Weight:
218.05
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing