
Dehydrocholic acid

Synonym(s):
(5β)-3,7,12-Trioxocholan-24-oic acid, 3,7,12-Trioxo-5β-cholanic acid, 5β-Cholanic acid-3,5,12-trione
Empirical Formula (Hill Notation):
C24H34O5
CAS Number:
Molecular Weight:
402.52
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing