
Chlorimuron ethyl

Synonym(s):
Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate
Empirical Formula (Hill Notation):
C15H15ClN4O6S
CAS Number:
Molecular Weight:
414.82
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2