
Bemotrizinol

Synonym(s):
2,4-Bis[4-(2-ethylhexyloxy)-2-hydroxyphenyl]-6-(4-methoxyphenyl)-1,3,5-triazine
Empirical Formula (Hill Notation):
C38H49N3O5
CAS Number:
Molecular Weight:
627.81
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3