
Ampicillin trihydrate

Synonym(s):
D-(−)-α-Aminobenzylpenicillin, Ampicillin trihydrate
Empirical Formula (Hill Notation):
C16H19N3O4S · 3H2O
CAS Number:
Molecular Weight:
403.45
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing