Acrolein-DNPH solution
Synonym(s):
Acrolein-2,4-dinitrophenylhydrazone solution
Empirical Formula (Hill Notation):
C9H8N4O4
CAS Number:
Molecular Weight:
236.18
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing