
Acitretin Related Compound B

Synonym(s):
Ethyl all-trans-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nonatetraenoate, Etretinate
Empirical Formula (Hill Notation):
C23H30O3
CAS Number:
Molecular Weight:
354.48
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2