Skip to Content
Merck
(3RS)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazin-7-one certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

(3RS)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazin-7-one

(3RS)-9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazin-7-one certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C17H20FN3O2
CAS Number:
Molecular Weight:
317.36

Products

Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Special Grade