Skip to Content
Merck
3-Methyl-2-buten-1-ol ≥98%, FG

3-Methyl-2-buten-1-ol

3-Methyl-2-buten-1-ol ≥98%, FG
Synonym(s):
3,3-Dimethylallyl alcohol, Prenol
Linear Formula:
(CH3)2C=CHCH2OH
CAS Number:
Molecular Weight:
86.13
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing