
2-Keto-D-gluconic acid hemicalcium salt hydrate

Synonym(s):
D-Arabino-2-hexulopyranosonic acid hemicalcium salt hydrate, Calcium 2-keto-D-gluconate
Linear Formula:
C6H9O7 · ½Ca · xH2O
CAS Number:
Molecular Weight:
213.17 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing