
14-Deoxy-11,14-didehydroandrographolide

Synonym(s):
14-Deoxy-11,12-didehydroandrographolide, 3-[(1E)-2-[(1R,4aS,5R,6R,8aR)-Decahydro-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethenyl]- 2(5H)-furanone, ent-(3β,11E)-3,19-Dihydroxy-8(17),11,13-labdatrien-16,15-olide
Empirical Formula (Hill Notation):
C20H28O4
CAS Number:
Molecular Weight:
332.43
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing