Merck
HomeProductsLabwarePCR Supplies & Equipment
PCR Supplies & Equipment
The polymerase chain reaction (PCR) is a powerful core molecular biology technique. It is an efficient and rapid in vitro method for enzymatic amplification of specific DNA or RNA sequences from nucleic acids of various sources. A simple PCR reaction consists of a set of synthetic oligonucleotide primers that flank the target DNA sequence, target DNA, a thermostable DNA polymerase and dNTPs. A repetitive series of cycles involving template denaturation, primer annealing, followed by extension of the annealed primers, yields tremendous amounts of DNA. Because the strands synthesized in one cycle serve as a template in the next, a million-fold increase in the DNA amount is achieved in just 20 cycles. From PCR microplates, plate covers, pipette tips, PCR tubes, and thermocyclers, explore our comprehensive offering of PCR consumables and equipment to support every step of your PCR workflow. 
Read more about
Products
Sort by Relevance
Showing 1-20 of 798
Change View
Showing 1-20 of 798
Compare
Product Number
Product Name
Product Description
Pricing
PCR Thermocyclers and Mini Cyclers for DNA Amplification
A thermal cycler (also known as a PCR machine or thermocycler) is a laboratory instrument that heats and cools samples in repetitive cycles to facilitate DNA or RNA amplification through the polymerase chain reaction. These machines have become commonplace in labs, although the introduction of mini or portable thermal cyclers has made it easier to perform PCR reactions in the field and classroom. Many thermal cyclers are programmable and allow users to save frequently used protocols. PCR typically requires 20 to 35 cycles comprising two to three temperature steps (denaturation, annealing, and extension). Combining speed, flexibility, and PCR optimization, the Eppendorf® Mastercycler Thermocyclers are suitable for enhanced PCR functions, like the 2D-Gradient and thorough documentation capabilities. Additionally, we also offer Benchmark Scientific TC models of thermocyclers that are designed for lower throughput needs, while maintaining the same functionality and a lower price point compared to larger units. 
Additionally, discover a variety of additional PCR plate seals, including eXTReme™ plate seals, ThermalSeal RTS™ plate seals, and AlumaSeal® PCR plate seals to accommodate your PCR workflow needs.
PCR plates, Strips and Films
With BRAND® PCR products you can protect your samples from contamination and evaporation. Cleanroom production, high-quality raw materials, and seamless quality controls by accredited laboratories ensure consistently high standards for cleanliness. Sealing films and covers made to fit plates and tubes significantly reduce evaporation loss and allow for reliable analyses, even with small sample volumes. BRAND® PCR consumable materials are always free from DNase, DNA, RNase, and endotoxins. We also offer Eppendorf® twin-tec PCR plates that combine the advantages of using both polypropylene and polycarbonate materials while the Corning® Thermowell PCR 96 well plates are suitable for the majority of thermocylers and ideal for high-throughput automation.
PCR Pipette Tips and Filter Tips
Explore a variety of reliable pipet tips, including market-leaders such as ART® pipet tips in our product selection table. In addition, BRAND® pipette tips and filter tips are manufactured in a cleanroom under the most modern production conditions and are automatically rack packed and packaged to ensure that the tips are of consistently high quality. BRAND® pipette tips and filter tips are manufactured with high-purity polypropylene, free from DiHEMDA, oleamide, DNA (< 40 fg), RNase (< 8.6 fg), endotoxins (< 1 pg) and ATP (< 1 fg). Additionally, we are proud to offer a portfolio of Eppendorf® products such as Eppendorf pipette tips that are used for liquid handling applications. They are useful for speed processing and avoiding cross-contamination. We offer a broad range of Eppendorf® pipette tips for niche applications according to the volume capacities, sterile and non-sterile demands as well as racking. Furthermore, Axygen consumables include not only pipette tips but also a variety of microplate options to match your specific application needs. 
Related Product Resources
  • Brochure: Reagents for PCR
    Roche has provided the gold standard in PCR reagents. Our advanced isolation, purification, and manufacturing techniques ensure that you receive not only the clearest, most reliable results, but also convenience and affordability to fit any laboratory budget.
  • Brochure: PCR
    Suitable for use with 96-well PCR plates as well as standard and low profile Thermo-Strips. Easy to apply without the aid of a capping tool. The flat surface of each cap facilitates sample labeling using a marker pen or adhesive stickers and large labeled end-tabs aid orientation and removal.
  • Tech Article: Complete Solutions for PCR Assay Development
    We offer a wide variety of Polymerase Chain Reaction (PCR) reagents, kits and tools to meet your experimental needs at any stage of development or manufacturing.
  • Protocol: PCR/qPCR/dPCR Assay Design
    The process of assay design is greatly facilitated by adoption of suitable design software. OligoArchitect provides two options for design support. The first is OligoArchitect Online, a software design tool with a wide range of options. If the design requires a specialized capability, the second option is to request the design via OligoArchitect Consultative, utilizing the assistance of our expert molecular biologists.
  • Protocol: FFPE Tissue
    Archived Formalin-fixed, Paraffin-embedded (FFPE) tissue samples are invaluable resources for profiling gene expression and studying a variety of diseases. Since the archived DNA is usually available in limited quantities, amplification of the samples is essential.
  • Quick Reference Guide: ProbeFinder
Page 1 of 2