Skip to Content
Merck
All Photos(1)

Key Documents

565306-U

Supelco

Ascentis® RP-Amide (5 µm) HPLC Columns

L × I.D. 25 cm × 2.1 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Ascentis® RP-Amide HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm

material

stainless steel column

Agency

suitable for USP L60

product line

Ascentis®

feature

endcapped

manufacturer/tradename

Ascentis®

packaging

1 ea of

extent of labeling

19.5% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

25 cm × 2.1 mm

surface area

450 m2/g

surface coverage

2.7 μmol/m2

impurities

<5 ppm metals

matrix

fully porous particle
silica gel high purity, spherical

matrix active group

amide, alkyl phase

particle size

5 μm

pore size

100 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

The Ascentis family of columns is the fourth generation of HPLC column technology from Supelco scientists. Ascentis columns are bonded on high purity, 100 Angstrom silica including 3, 5, and 10 micron particle size. Columns are designed for small molecule applications and are scalable from micro columns (1.0 mm I.D.) to preparative dimensions (50 mm I.D.). The family includes C18, C8, Phenyl, Si and embedded polar group phase, RP-Amide.

Ascentis RP-Amide is a new generation ultra low bleed, embedded polar group (EPG) phase that provides orthogonal selectivity and increased resolution for HPLC and LC-MS analysis of polar compounds. The Ascentis RP-Amide is the first choice in embedded polar group HPLC phases.

Features and Benefits

  • Excellent retention and peak shape for polar compounds
  • 100% aqueous compatibility
  • Ultra low bleed, LC-MS compatible
  • Unique selectivity

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jakub Fibigr et al.
Journal of pharmaceutical and biomedical analysis, 151, 291-300 (2018-02-08)
The presented work describes the development and validation of a rapid UHPLC-UV method using a fused core particle column with an RP-Amide stationary phase for the separation and quantitative analysis of caffeoylquinic and di-caffeoylquinic acids in green coffee extracts. Three
Rihana Parveen Shaik et al.
Journal of pharmaceutical analysis, 3(6), 489-499 (2013-12-01)
Liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was used for simultaneous quantification of tolterodine and its metabolite 5-hydroxy methyl tolterodine in rat plasma. Tolterodine-d6 and 5-hydroxy methyl tolterodine-d14 were used as internal standards (IS). Chromatographic separation was performed on Ascentis Express

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service