Skip to Content
Merck
All Photos(2)

Key Documents

21822-U

Supelco

Empty Tube for Thermal Desorption

greener alternative

stainless steel (includes screens), O.D. x I.D. x L 1/4 in. × 3 1/2, pkg of 10 ea

Synonym(s):

empty stainless Thermal Desorption Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

product name

Empty Tube for Thermal Desorption, stainless steel TD tube (includes screens), O.D. × L 1/4 in. × 3 1/2, pkg of 10 ea

material

stainless steel TD tube (includes screens)

packaging

pkg of 10 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Empty Tube for Thermal Desorption features an etched unique number for sample identification and is packed with high quality adsorbents, preconditioned and sealed in a TDS3 Storage Container.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Application


  • Performance test of a sorbent tube sampler with respect to analyte loss in collecting biogenic volatile organic compounds: This research examines the efficiency of Empty Tube for Thermal Desorption when used in the collection of biogenic volatile organic compounds (VOCs). Highlighting its critical role in minimizing analyte loss, this study underscores the tube′s importance in environmental sampling and analysis. It also touches on applications across environmental monitoring and analytical chemistry, illustrating its utility in ensuring accurate data collection and integrity in laboratory settings (Ullah & Kim, 2015).


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service