Skip to Content
Merck
All Photos(1)

Key Documents

1.08387

Millipore

Tris Base

solid, TRIS LAB

Synonym(s):

Tris Base, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-064-4
NACRES:
NA.05

product name

Tris(hydroxymethyl)aminomethane, TRIS LAB

vapor pressure

0.0267 hPa ( 20 °C)

Quality Level

form

solid

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rat)

pH

10.2-10.6 (20 °C, 6 g/L in H2O)

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)
219-220 °C/13.3 hPa

mp

167-172 °C (lit.)

solubility

678 g/L

density

1.35 g/cm3 at 23 °C

bulk density

840 kg/m3

storage temp.

15-25°C

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

TRIS LAB

Application

Tris(hydroxymethyl)aminomethane TRIS LAB. CAS 77-86-1, EC 201-064-4, chemical formula H₂NC(CH₂OH)₃.

Analysis Note

Assay (acidimetric, calc. on anhydrous substance)99.8 - 100.1 %Identity (IR-spectrum)conformsIn water insoluble matter≤ 0.005 %Absorbance (290 nm; 40 %; 1 cm; water)≤ 0.2Chloride (Cl)≤ 0.001 %Sulfate (SO₄)≤ 0.005 %Heavy metals (as Pb)≤ 0.0005 %Fe (Iron)≤ 0.0005 %Sulfated ash≤ 0.05 %Water (according to Karl Fischer)≤ 0.5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service