Skip to Content
Merck
All Photos(1)

Key Documents

Z676195

Plum saline eyewash refill

pkg of 500 mL

Synonym(s):

Saline eyewash solution, Sterile saline rinse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131905
NACRES:
NB.35
Pricing and availability is not currently available.

packaging

pkg of 500 mL

manufacturer/tradename

Plum

General description

A bottle containing a 0.9% sterile sodium chloride solution. The bottle can be used alone or as a refill for eyewash stations. The bottle is provided with an ergonomic eyecup, dust cap and clear instructions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service