Key Documents
Z609676
Bohlender screw caps
GL 32 thread
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
manufacturer/tradename
Bohlender
availability
not available in USA and Canada
fitting
GL 32 thread
General description
Made of PPS these screw caps with virgin PTFE membrane gaskets are resistant to high mechanical stresses - even at temperatures upto 250 °C. The gasket consists of PTFE membrane with a multidirectional orientated fibre pattern. Medium will only be exposed to PTFE.
Legal Information
Bohlender is a trademark of Bohlender GmbH