Key Documents
Z319260
Sigma-Aldrich
Drying tube sublimation insert for Büchi® models B-580 & B-585 ovens
borosilicate glass, pkg of 1 ea
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
material
borosilicate glass
packaging
pkg of 1 ea
manufacturer/tradename
Büchi 037133
General description
As an alternative use, the Buchi B-585 glass drying oven (Z563080 or Z616818) has a sublimation drying tube insert where substances can be physically cleansed via sublimation. A solid matter passes directly into its gaseous state and is resublimated on a cold surface. The sublimation accessory is inserted into the drying oven and cooled with water or another cooling medium.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG