566802
Toluene-4-sulfonic acid dibenzyl-α-ylidenehydrazone
97%
Synonym(s):
p-Toluenesulfonic acid (1,2-diphenylethylidene)hydrazide
About This Item
Recommended Products
Assay
97%
form
solid
mp
139-143 °C (lit.)
SMILES string
Cc1ccc(cc1)S(=O)(=O)N\N=C(/Cc2ccccc2)c3ccccc3
InChI
1S/C21H20N2O2S/c1-17-12-14-20(15-13-17)26(24,25)23-22-21(19-10-6-3-7-11-19)16-18-8-4-2-5-9-18/h2-15,23H,16H2,1H3/b22-21+
InChI key
INMYHVHTZKDKDK-QURGRASLSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service