Skip to Content
Merck
All Photos(1)

Key Documents

306894

Sigma-Aldrich

3-Hexyne

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C≡CC2H5
CAS Number:
Molecular Weight:
82.14
Beilstein:
1731158
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

186 mmHg ( 37.7 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.411 (lit.)

bp

81-82 °C (lit.)

density

0.723 g/mL at 25 °C (lit.)

SMILES string

CCC#CCC

InChI

1S/C6H10/c1-3-5-6-4-2/h3-4H2,1-2H3

InChI key

DQQNMIPXXNPGCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Hexyne was used in the synthesis of fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety by reacting with ruthenacycles. It was also used in the preparation of [4+2] cycloaddition product by reacting with borole.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

6.8 °F - closed cup

Flash Point(C)

-14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Ge et al.
Journal of the American Chemical Society, 136(1), 68-71 (2013-12-21)
Bis(trimethylsilylethynyl)diphenylaminoborane was reacted with the strong Lewis acid B(C6F5)3 at ambient temperature to give the borole 9 admixed with a small amount of its thermal follow-up product 12. Compound 9 was subsequently stabilized by adduct formation with pyridine (10). Treatment
Luciano Cuesta et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(47), 15178-15189 (2012-09-29)
Novel cycloruthenated complexes 2 a-c, 4 a-c, and 6 a, b based on heteroaromatic cores have been synthesized by reaction of a series of heterocycle-based imines with [{RuCl(η(6)-p-cymene)}(2)(μ-Cl)(2)] and Cu(OAc)(2). This approach has proved efficient for the cyclometalation of thiophene, benzothiophene, furan, benzofuran, pyrrole
He Nan et al.
Journal of chromatography. A, 1523, 316-320 (2017-06-26)
Silver ion or argentation chromatography utilizes stationary phases containing silver ions for the separation of unsaturated compounds. In this study, a mixed-ligand silver-based ionic liquid (IL) was evaluated for the first time as a gas chromatographic (GC) stationary phase for
Davide Albani et al.
Nature communications, 9(1), 2634-2634 (2018-07-08)
Ensemble control has been intensively pursued for decades to identify sustainable alternatives to the Lindlar catalyst (PdPb/CaCO3) applied for the partial hydrogenation of alkynes in industrial organic synthesis. Although the geometric and electronic requirements are known, a literature survey illustrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service