Merck
  • The Hypothalamic Inflammatory/Gliosis Response to Neonatal Overnutrition Is Sex and Age Dependent.
The Hypothalamic Inflammatory/Gliosis Response to Neonatal Overnutrition Is Sex and Age Dependent.
Endocrinology (2017-10-28)
Pilar Argente-Arizón, Francisca Díaz, Purificación Ros, Vicente Barrios, Manuel Tena-Sempere, Luis Miguel García-Segura, Jesús Argente, Julie A Chowen
ABSTRACT
Astrocytes participate in both physiological and pathophysiological responses to metabolic and nutrient signals. Although most studies have focused on the astrocytic response to weight gain due to high-fat/high-carbohydrate intake, surplus intake of a balanced diet also induces excess weight gain. We have accessed the effects of neonatal overnutrition, which has both age- and sex-dependent effects on weight gain, on hypothalamic inflammation/gliosis. Although both male and female Wistar rats accumulate excessive fat mass as early as postnatal day (PND) 10 with neonatal overnutrition, no increase in hypothalamic cytokine levels, markers of astrocytes or microglia, or inflammatory signaling pathways were observed. At PND 50, no effect of neonatal overnutriton was found in either sex, whereas at PND 150, males again weighed significantly more than their controls, and this was coincident with an increase in markers of inflammation and astrogliosis in the hypothalamus. Circulating triglycerides and free fatty acids were also elevated in these males, but not in females or in either sex at PND 10. Thus, the effects of fatty acids and estrogens on astrocytes in vitro were analyzed. Our results indicate that changes in circulating fatty acid levels may be involved in the induction of hypothalamic inflammation/gliosis in excess weight gain, even on a normal diet, and that estrogens could participate in the protection of females from these processes. In conclusion, the interaction of developmental influences, dietary composition, age, and sex determines the central inflammatory response and the associated long-term outcomes of excess weight gain.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Bovine Serum Albumin solution, 30% in saline, fatty acid free, aseptically filled
Sigma-Aldrich
Monoclonal Anti-PTEN antibody produced in mouse, ~1 mg/mL, purified immunoglobulin, clone PTN-18, buffered aqueous solution