Merck
  • Molecular Characteristics and Serodiagnostic Potential of Dihydrofolate Reductase from Echinococcus granulosus.
Molecular Characteristics and Serodiagnostic Potential of Dihydrofolate Reductase from Echinococcus granulosus.
Scientific reports (2017-04-02)
Xingju Song, Dandan Hu, Min Yan, Yu Wang, Ning Wang, Xiaobin Gu, Guangyou Yang
ABSTRACT
The larval stage of Echinococcus granulosus causes cystic echinococcosis (CE), a neglected tropical disease that leads to morbidity and mortality in humans and livestock worldwide. Here, we identified and characterized dihydrofolate reductase (Eg-DHFR) from E. granulosus, and evaluated its potential as a diagnostic antigen for sheep CE. Comparison between mammalian (host) DHFR and Eg-DHFR indicates that 45.7% of the 35 active site residues are different. Immunolocalisation analysis showed that native Eg-DHFR was widely distributed in all life-cycle stages of E. granulosus. Recombinant Eg-DHFR (rEg-DHFR) showed typical DHFR enzymatic parameters towards substrate, and was very sensitive to inhibition by methotrexate (IC
Sigma-Aldrich
Dihydrofolic acid, ≥90%