Merck
  • Inhibition of platelet-derived growth factor-BB-induced receptor activation and fibroblast migration by hyaluronan activation of CD44.
Inhibition of platelet-derived growth factor-BB-induced receptor activation and fibroblast migration by hyaluronan activation of CD44.
The Journal of biological chemistry (2006-07-01)
Lingli Li, Carl-Henrik Heldin, Paraskevi Heldin
ABSTRACT
The extracellular matrix molecule hyaluronan was found to suppress platelet-derived growth factor (PDGF) beta-receptor activation and PDGF-BB-induced migration of primary human dermal fibroblasts. The suppressive effect of hyaluronan was neutralized by a monoclonal antibody that specifically inhibits hyaluronan binding to its receptor CD44. Moreover, co-immunoprecipitation experiments showed that the PDGF beta-receptor and CD44 can form a complex. Interestingly, the inhibitory effect of hyaluronan on PDGF beta-receptor activation was not seen in the presence of the tyrosine phosphatase inhibitor pervanadate. Our observations suggest that hyaluronan suppresses PDGF beta-receptor activation by recruiting a CD44-associated tyrosine phosphatase to the receptor.
Sigma-Aldrich
3, 3′,5 ,5′-Tetramethylbenzidine Liquid Substrate, Supersensitive, for ELISA, ready to use solution
Sigma-Aldrich
Biotinamidohexanoic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt, ≥90% (TLC), powder