Merck
  • Interleukin-18 produced by bone marrow-derived stromal cells supports T-cell acute leukaemia progression.
Interleukin-18 produced by bone marrow-derived stromal cells supports T-cell acute leukaemia progression.
EMBO molecular medicine (2014-04-30)
Benjamin Uzan, Sandrine Poglio, Bastien Gerby, Ching-Lien Wu, Julia Gross, Florence Armstrong, Julien Calvo, Xavier Cahu, Caroline Deswarte, Florent Dumont, Diana Passaro, Corinne Besnard-Guérin, Thierry Leblanc, André Baruchel, Judith Landman-Parker, Paola Ballerini, Véronique Baud, Jacques Ghysdael, Frédéric Baleydier, Francoise Porteu, Francoise Pflumio
ABSTRACT
Development of novel therapies is critical for T-cell acute leukaemia (T-ALL). Here, we investigated the effect of inhibiting the MAPK/MEK/ERK pathway on T-ALL cell growth. Unexpectedly, MEK inhibitors (MEKi) enhanced growth of 70% of human T-ALL cell samples cultured on stromal cells independently of NOTCH activation and maintained their ability to propagate in vivo. Similar results were obtained when T-ALL cells were cultured with ERK1/2-knockdown stromal cells or with conditioned medium from MEKi-treated stromal cells. Microarray analysis identified interleukin 18 (IL-18) as transcriptionally up-regulated in MEKi-treated MS5 cells. Recombinant IL-18 promoted T-ALL growth in vitro, whereas the loss of function of IL-18 receptor in T-ALL blast cells decreased blast proliferation in vitro and in NSG mice. The NFKB pathway that is downstream to IL-18R was activated by IL-18 in blast cells. IL-18 circulating levels were increased in T-ALL-xenografted mice and also in T-ALL patients in comparison with controls. This study uncovers a novel role of the pro-inflammatory cytokine IL-18 and outlines the microenvironment involvement in human T-ALL development.
Sigma-Aldrich
Human IL-18 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IL-18 BPa  ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse Il18 / Interleukin-18 ELISA Kit, for serum, plasma and cell culture supernatants