Merck
  • Neuroendocrine differentiation in pancreatic duct carcinoma special emphasis on duct-endocrine cell carcinoma of the pancreas.
Neuroendocrine differentiation in pancreatic duct carcinoma special emphasis on duct-endocrine cell carcinoma of the pancreas.
Pathology, research and practice (1996-09-01)
T Kamisawa, M Fukayama, I Tabata, T Isawa, K Tsuruta, A Okamoto, Koike
ABSTRACT
To evaluate the significance of neuroendocrine differentiation in duct carcinoma of the pancreas, we investigated 79 pancreatic carcinomas, applying histochemistry and immunohistochemistry (chromogranin A, Leu-7, synaptophysin and neuron-specific enolase (NSE), and correlated the morphologic differentiation pattern with clinicopathological characteristics. There were two types of neuroendocrine differentiation: scattered (n = 23) and diffuse (n = 3). The scattered type of pancreatic duct carcinoma contained scattered endocrine cells amounting to less than 10% of the neoplastic cells and was seen more frequently in well-differentiated carcinomas. There was no characteristic clinical feature found in the scattered type when compared with the tumors devoid of endocrine cells (n = 53). In contrast, the diffuse type showed diffuse immunostaining with NSE and synaptophysin in tumor cells and dense core granules ultrastructurally. These tumors showed a greater hypervascularity in angiography (p < 0.01) and the patients had relatively longer survival (33.3 months, p < 0.05) than unresectable cases of other histological types of pancreatic cancer. Two types of neuroendocrine differentiation (scattered and diffuse) existed in pancreatic ductal carcinoma. The diffuse type (Duct-Endocrine Cell Carcinoma of the Pancreas) showed synchronous duct and endocrine differentiation and particular clinical features.
Sigma-Aldrich
Synaptophysin (EP158) Rabbit Monoclonal Primary Antibody