Merck
  • MS disease activity in RESTORE: a randomized 24-week natalizumab treatment interruption study.
MS disease activity in RESTORE: a randomized 24-week natalizumab treatment interruption study.
Neurology (2014-04-01)
Robert J Fox, Bruce A C Cree, Jerome De Sèze, Ralf Gold, Hans-Peter Hartung, Douglas Jeffery, Ludwig Kappos, Michael Kaufman, Xavier Montalbán, Bianca Weinstock-Guttman, Britt Anderson, Amy Natarajan, Barry Ticho, Petra Duda
ABSTRACT
RESTORE was a randomized, partially placebo-controlled exploratory study evaluating multiple sclerosis (MS) disease activity during a 24-week interruption of natalizumab. Eligible patients were relapse-free through the prior year on natalizumab and had no gadolinium-enhancing lesions on screening brain MRI. Patients were randomized 1:1:2 to continue natalizumab, to switch to placebo, or to receive alternative immunomodulatory therapy (other therapies: IM interferon β-1a [IM IFN-β-1a], glatiramer acetate [GA], or methylprednisolone [MP]). During the 24-week randomized treatment period, patients underwent clinical and MRI assessments every 4 weeks. Patients (n = 175) were randomized to natalizumab (n = 45), placebo (n = 42), or other therapies (n = 88: IM IFN-β-1a, n = 17; GA, n = 17; MP, n = 54). Of 167 patients evaluable for efficacy, 49 (29%) had MRI disease activity recurrence: 0/45 (0%) natalizumab, 19/41 (46%) placebo, 1/14 (7%) IM IFN-β-1a, 8/15 (53%) GA, and 21/52 (40%) MP. Relapse occurred in 4% of natalizumab patients and in 15%-29% of patients in the other treatment arms. MRI disease activity recurred starting at 12 weeks (n = 3 at week 12) while relapses were reported as early as 4-8 weeks (n = 2 in weeks 4-8) after the last natalizumab dose. Overall, 50/167 patients (30%), all in placebo or other-therapies groups, restarted natalizumab early because of disease activity. MRI and clinical disease activity recurred in some patients during natalizumab interruption, despite use of other therapies. This study provides Class II evidence that for patients with MS taking natalizumab who are relapse-free for 1 year, stopping natalizumab increases the risk of MS relapse or MRI disease activity as compared with continuing natalizumab.
Product Description
USP
Methylprednisolone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
6α-Methylprednisolone, ≥98%
Methylprednisolone, European Pharmacopoeia (EP) Reference Standard