Merck
  • Spectroscopic, electronic structure and natural bond orbital analysis of o-fluoronitrobenzene and p-fluoronitrobenzene: a comparative study.
Spectroscopic, electronic structure and natural bond orbital analysis of o-fluoronitrobenzene and p-fluoronitrobenzene: a comparative study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-10-14)
V Arjunan, S Thillai Govindaraja, S Sakiladevi, M Kalaivani, S Mohan
ABSTRACT
Experimental FTIR, FT-Raman and FT-NMR spectroscopic studies of o-fluoronitrobenzene and p-fluoronitrobenzene have been carried out. A detailed quantum chemical calculations have been performed using DFT/B3LYP method with 6-311++G** and 6-31G** basis sets. Complete vibrational analyses of the compounds were performed. The temperature dependence of thermodynamic properties has been analysed. The atomic charges, electronic exchange interaction and charge delocalisation of the molecule have been performed by natural bond orbital (NBO) analysis. Molecular electrostatic surface potential (MESP), total electron density distribution and frontier molecular orbitals (FMOs) are constructed at B3LYP/6-311++G** level to understand the electronic properties. The charge density distribution and site of chemical reactivity of the molecules have been obtained by mapping electron density isosurface with electrostatic potential surfaces (ESP). The electronic properties, HOMO and LUMO energies were measured by time-dependent TD-DFT approach. (1)H and (13)C NMR spectra were recorded and (1)H and (13)C nuclear magnetic resonance chemical shifts of the molecule were calculated. The (1)H and (13)C nuclear magnetic resonance (NMR) chemical shifts of the molecules in chloroform solvent and in gas phase were calculated by using the Gauge-Independent Atomic Orbital (GIAO) method and are found to be in good agreement with experimental values. The theoretical parameters obtained at B3LYP levels have been compared with the experimental values.
Sigma-Aldrich
1-Fluoro-2-nitrobenzene, 99%