Merck
  • Inhibitory effects of (-)-α-bisabolol on LPS-induced inflammatory response in RAW264.7 macrophages.
Inhibitory effects of (-)-α-bisabolol on LPS-induced inflammatory response in RAW264.7 macrophages.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-07-21)
Seungbeom Kim, Eunsun Jung, Jang-Hyun Kim, Young-Ho Park, Jongsung Lee, Deokhoon Park
ABSTRACT
Although (-)-α-bisabolol, a natural monocyclic sesquiterpene alcohol, is often used as a cosmetic soothing supplement, little is known about its mechanisms of anti-inflammatory effects. Therefore, this study was designed to investigate anti-inflammatory effects of (-)-α-bisabolol and its mechanisms of action. In this study, we found that (-)-α-bisabolol inhibited lipopolysaccharide (LPS)-induced production of nitric oxide (NO) and prostaglandin E(2) (PGE(2)) in RAW264.7 cells. In addition, expression of cyclooxygenase-2 (COX-2) and inducible nitric oxide synthase (iNOS) genes was reduced, as evidenced by Western blot and luciferase reporter assays for COX-2 and iNOS. To assess the mechanism of the anti-inflammatory property of (-)-α-bisabolol, its effects on the activity of AP-1 and NF-κB promoters were examined. LPS-induced activation of AP-1 and NF-κB promoters was significantly reduced by (-)-α-bisabolol. Consistently, (-)-α-bisabolol reduced LPS-induced phosphorylation of IκBα. In addition, while LPS-induced phosphorylation of ERK and p38 was attenuated by (-)-α-bisabolol, significant changes in the level of phosphorylated JNK were not observed. Our results indicate that (-)-α-bisabolol exerts anti-inflammatory effects by downregulating expression of iNOS and COX-2 genes through inhibition of NF-κB and AP-1 (ERK and p38) signaling.
Sigma-Aldrich
(−)-α-Bisabolol, ≥93% (GC)
Supelco
(−)-α-Bisabolol, analytical standard
(−)-α-Bisabolol, primary reference standard