Merck
  • Microbial biodegradation of microcystin-RR by bacterium Sphingopyxis sp. USTB-05.
Microbial biodegradation of microcystin-RR by bacterium Sphingopyxis sp. USTB-05.
Journal of environmental sciences (China) (2010-04-20)
Mulan Zhang, Gang Pan, Hai Yan
ABSTRACT
A strain, USTB-05, isolated from Lake Dianchi, China, degraded the cyanobacterial toxin microcystin-RR (MC-RR) at the rate of 16.7 mg/L per day. Analysis of 16S rDNA sequence showed that the strain was Sphingopyxis sp. Enzymatic degradation pathways for MC-RR by Sphingopyxis sp. USTB-05 were identified. Adda-Arg peptide bond of MC-RR was cleaved and then a hydrogen and a hydroxyl were combined onto the NH2 group of Adda and the carboxyl group of arginine to form a linear molecule as intermediate product within the first few hours. Then, through dehydration reaction, two hydrogen of amino group on arginine reacted with adjacent hydroxyl on carbon to form a linear MC-RR with two small peptide rings as the final product after 24 hr. These biodegradation pathways were different from those reported for other strains, implying that MC-RR may undergo different transformations and different products were formed due to various bacteria in natural lakes and reservoirs.
Product Description
Supelco
Microcystin-RR solution, 10 μg/mL in methanol, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
1 Estimated to ship on 16 February 2025
CHF 249.00
50 μg
1 Estimated to ship on 16 February 2025
CHF 443.00