Merck
  • Crystal structures of Escherichia coli gamma-glutamyltranspeptidase in complex with azaserine and acivicin: novel mechanistic implication for inhibition by glutamine antagonists.
Crystal structures of Escherichia coli gamma-glutamyltranspeptidase in complex with azaserine and acivicin: novel mechanistic implication for inhibition by glutamine antagonists.
Journal of molecular biology (2008-06-17)
Kei Wada, Jun Hiratake, Machiko Irie, Toshihiro Okada, Chiaki Yamada, Hidehiko Kumagai, Hideyuki Suzuki, Keiichi Fukuyama
ABSTRACT
gamma-Glutamyltranspeptidase (GGT) catalyzes the cleavage of such gamma-glutamyl compounds as glutathione, and the transfer of their gamma-glutamyl group to water or to other amino acids and peptides. GGT is involved in a number of biological phenomena such as drug resistance and metastasis of cancer cells by detoxification of xenobiotics. Azaserine and acivicin are classical and irreversible inhibitors of GGT, but their binding sites and the inhibition mechanisms remain to be defined. We have determined the crystal structures of GGT from Escherichia coli in complex with azaserine and acivicin at 1.65 A resolution. Both inhibitors are bound to GGT at its substrate-binding pocket in a manner similar to that observed previously with the gamma-glutamyl-enzyme intermediate. They form a covalent bond with the O(gamma) atom of Thr391, the catalytic residue of GGT. Their alpha-carboxy and alpha-amino groups are recognized by extensive hydrogen bonding and charge interactions with the residues that are conserved among GGT orthologs. The two amido nitrogen atoms of Gly483 and Gly484, which form the oxyanion hole, interact with the inhibitors directly or via a water molecule. Notably, in the azaserine complex the carbon atom that forms a covalent bond with Thr391 is sp(3)-hybridized, suggesting that the carbonyl of azaserine is attacked by Thr391 to form a tetrahedral intermediate, which is stabilized by the oxyanion hole. Furthermore, when acivicin is bound to GGT, a migration of the single and double bonds occurs in its dihydroisoxazole ring. The structural characteristics presented here imply that the unprecedented binding modes of azaserine and acivicin are conserved in all GGTs from bacteria to mammals and give a new insight into the inhibition mechanism of glutamine amidotransferases by these glutamine antagonists.
Sigma-Aldrich
Azaserine, Hybri-Max, γ-irradiated, 50x, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Azaserine, ≥98% (TLC)