Merck
  • Liquid chromatographic analysis of propranolol enantiomers in human blood using precolumn derivatization with (+)-1-(9-fluorenyl)ethyl chloroformate.
Liquid chromatographic analysis of propranolol enantiomers in human blood using precolumn derivatization with (+)-1-(9-fluorenyl)ethyl chloroformate.
Journal of chromatography (1991-10-04)
A Roux, G Blanchot, A Baglin, B Flouvat
ABSTRACT
A method for the determination of the R-(+) and S-(-) enantiomers of propranolol in blood was developed. After extraction with heptane-isopentanol and derivatization with (+)-1-(9-fluorenyl)ethyl chloroformate, excess reagent was removed using solid-phase extraction. The enantiomers were separated on an achiral, reversed-phase, radially compressed column, and detected by fluorescence with excitation and emission wavelengths of 260 and 340 nm, respectively. The limit of quantification was 0.5 ng/ml. This method was used for pharmacokinetic analysis of propranolol enantiomers after administration of immediate-release (80 mg) or sustained-release (160 mg) racemic propranolol.
Product Description
Supelco
(+)-1-(9-Fluorenyl)ethyl chloroformate solution, ≥18 mM in acetone, for chiral derivatization, LiChropur
Sigma-Aldrich
(+)-1-(9-Fluorenyl)ethyl chloroformate solution, 18 mM in acetone, for chiral derivatization