Merck
  • Comprehensive authentication of (E)-alpha(beta)-ionone from raspberries, using constant flow MDGC-C/P-IRMS and enantio-MDGC-MS.
Comprehensive authentication of (E)-alpha(beta)-ionone from raspberries, using constant flow MDGC-C/P-IRMS and enantio-MDGC-MS.
Journal of agricultural and food chemistry (2005-02-17)
Sabine Sewenig, Dino Bullinger, Uwe Hener, Armin Mosandl
ABSTRACT
A new coupling system of GC-GC, connected via a Multi Column Switching Device MCS2 for measuring isotope ratios, is introduced. By means of several standard substances the precise and accurate measurement of isotopic values is proved. First applications concerning the authentication of raspberry aroma compounds are established. Consequently, the combination of constant flow multidimensional gas chromatography-combustion/pyrolysis-isotope ratio mass spectrometry (MDGC-C/P-IRMS) is applied to the authenticity assessment of (E)-alpha(beta)-ionone from six different raspberry cultivars. Furthermore, 12 commercially available raspberry products and samples of (E)-alpha(beta)-ionone, some declared to be natural, are investigated. delta(2)Eta(V)(-)(SMOW) and delta(13)C(V)(-)(PDB) values of (E)-alpha(beta)-ionone are determined, and characteristic authenticity ranges were concluded from raspberries by correlation of both delta(2)Eta(V)(-)(SMOW) and delta(13)C( V)(-)(PDB) values. The results are correlated with the determination of enantiomeric purities of (E)-alpha-ionone, using stir bar sorptive extraction enantio-multidimensional gas chromatography mass spectrometry (SBSE-enantio-MDGC-MS).
Sigma-Aldrich
α-Ionone, technical grade, 90%
Sigma-Aldrich
α-Ionone, ≥90%, stabilized