Merck
  • Structure of hydroxynitrile lyase from Manihot esculenta in complex with substrates acetone and chloroacetone: implications for the mechanism of cyanogenesis.
Structure of hydroxynitrile lyase from Manihot esculenta in complex with substrates acetone and chloroacetone: implications for the mechanism of cyanogenesis.
Acta crystallographica. Section D, Biological crystallography (2001-02-15)
H Lauble, S Förster, B Miehlich, H Wajant, F Effenberger
ABSTRACT
The crystal structures of hydroxynitrile lyase from Manihot esculenta (MeHNL) complexed with the native substrate acetone and substrate analogue chloroacetone have been determined and refined at 2.2 A resolution. The substrates are positioned in the active site by hydrogen-bond interactions of the carbonyl O atom with Thr11 OG, Ser80 OG and, to a lesser extent, Cys81 SG. These studies support a mechanism for cyanogenesis as well as for the stereospecific MeHNL-catalyzed formation of (S)-cyanohydrins, which closely resembles the base-catalyzed chemical reaction of HCN with carbonyl compounds.
Sigma-Aldrich
Chloroacetone, 95%
Sigma-Aldrich
Chloroacetone, Wacker Chemie AG, ≥96.0% (GC)