Merck
  • Vildagliptin ameliorates oxidative stress and pancreatic beta cell destruction in type 1 diabetic rats.
Vildagliptin ameliorates oxidative stress and pancreatic beta cell destruction in type 1 diabetic rats.
Archives of medical research (2013-03-26)
Danielle de Lima Ávila, Glaucy Rodrigues de Araújo, Maisa Silva, Pedro Henrique de Amorim Miranda, Mirla Fiuza Diniz, Maria Lúcia Pedrosa, Marcelo Eustáquio Silva, Wanderson Geraldo de Lima, Daniela Caldeira Costa
ABSTRACT
It is believed that oxidative stress plays a role in the pathogenesis of diabetes mellitus. Several strategies have been developed with the objective of minimizing diabetic complications. Among these, inhibitors of dipeptidyl peptidase-IV (DPP-IV), which act by blocking degradation of incretin hormones, glucagon-like peptide hormone (GLP-1) and glucose-dependent insulinotropic polypeptide (GIP), have been the focus of many studies. It is known that, among the effects of incretins, we highlight its insulinotropic and cytoprotective effects on pancreatic β-cells. The objective of this study was to evaluate the possible protective effects of treatment with vildagliptin, a DPP-IV inhibitor, in β-cells in an experimental model of type 1 diabetes induced by streptozotocin (STZ). Rats were treated for 4 weeks with vildagliptin at concentrations of 5 and 10 mg/kg. In order to observe the pancreatic damage and the possible protective effects of vildagliptin treatment, we measured stress markers TBARS and protein carbonyl, antioxidant enzymes SOD and catalase, and analyzed pancreatic histology. The treatment was effective in modulating stress in pancreatic tissue, both by reducing levels of stress markers as well as by increasing activity of SOD and catalase. After analyzing the pancreatic histology, we found that vildagliptin was also able to preserve islets and pancreatic β-cells, especially at the concentration of 5 mg/kg. Thus, our results suggest that vildagliptin ameliorates oxidative stress and pancreatic beta cell destruction in type 1 diabetic rats. However, to evaluate the real potential of this medication in type 1 diabetes, further studies are needed.
Sigma-Aldrich
Dipeptidyl Peptidase IV human, recombinant, expressed in baculovirus infected Sf9 cells, pkg of ≥1.0 units/vial, ≥10 units/mg protein
Sigma-Aldrich
Adamantane, ≥99%
Sigma-Aldrich
Dipeptidyl Peptidase IV human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein