- A TGF-β-responsive enhancer regulates SRC expression and epithelial-mesenchymal transition-associated cell migration.
A TGF-β-responsive enhancer regulates SRC expression and epithelial-mesenchymal transition-associated cell migration.
The non-receptor tyrosine kinase SRC is overexpressed and/or hyperactivated in various human cancers, and facilitates cancer progression by promoting invasion and metastasis. However, the mechanisms underlying SRC upregulation are poorly understood. In this study, we demonstrate that transforming growth factor-β (TGF-β) induces SRC expression at the transcriptional level by activating an intragenic the SRC enhancer. In the human breast epithelial cell line MCF10A, TGF-β1 stimulation upregulated one of the SRC promotors, the 1A promoter, resulting in increased SRC mRNA and protein levels. Chromatin immunoprecipitation (ChIP)-sequencing analysis revealed that the SMAD complex is recruited to three enhancer regions ∼15 kb upstream and downstream of the SRC promoter, and one of them is capable of activating the SRC promoter in response to TGF-β. JUN, a member of the activator protein (AP)-1 family, localises to the enhancer and regulates TGF-β-induced SRC expression. Furthermore, TGF-β-induced SRC upregulation plays a crucial role in epithelial-mesenchymal transition (EMT)-associated cell migration by activating the SRC-focal adhesion kinase (FAK) circuit. Overall, these results suggest that TGF-β-induced SRC upregulation promotes cancer cell invasion and metastasis in a subset of human malignancies.