- SARS-CoV-2 proteome microarray for COVID-19 patient sera profiling.
SARS-CoV-2 proteome microarray for COVID-19 patient sera profiling.
STAR protocols (2022-03-22)
Dan-Yun Lai, He-Wei Jiang, Yang Li, Hai-Nan Zhang, Sheng-Ce Tao
PMID35310073
ABSTRACT
The immunogenicity of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) proteome is largely unknown. Here we describe a protocol for analyzing sera samples with SARS-CoV-2 proteome microarray. The proteins were expressed by either E. coli expression system or eukaryotic cell expression systems and obtained by affinity purification. The protocol includes microarray fabricating and sera profiling, which will be used to build an antibody response landscape for IgG and IgM. The protocol may help to facilitate a deeper understanding of immunity related to SARS-CoV-2. For complete details on the use and execution of this protocol, please refer to Li et al. (2021c).
Sigma-Aldrich
Anti-Human IgG (Fc specific) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgM (μ-chain specific) antibody produced in goat, affinity isolated antibody, buffered aqueous solution