Merck
  • Implementation of a Physical Activity Program Protocol in Schoolchildren: Effects on the Endocrine Adipose Tissue and Cognitive Functions.
Implementation of a Physical Activity Program Protocol in Schoolchildren: Effects on the Endocrine Adipose Tissue and Cognitive Functions.
Frontiers in nutrition (2021-11-09)
Javier Diaz-Castro, Jose Eulogio Garcia-Vega, Julio J Ochoa, María Puche-Juarez, Juan M Toledano, Jorge Moreno-Fernandez
ABSTRACT
Practicing exercise is one of the best strategies to promote well-being and quality of life, however physical activity in schoolchildren and adolescents is developed in an unpredictable, intermittent way and in short periods. There are relatively few intervention studies investigating the role of physical activity in schoolchildren endocrine function of adipose tissue and cognitive function. One hundred and three boys, divided into two groups: control (n = 51, did not perform additional physical activity) and exercise (n = 52, performed vigorous physical activity after the regular school classes). The exercise group, developed a 6 months physical activity protocol delivered by the physical education teacher during the second semester of the academic course (6 months). Body composition measurements, adherence to the Mediterranean diet, nutritional intake, hematological and biochemical parameters, endocrine function of the adipose tissue and biomarkers of brain molecular function were assessed at enrolment and after 6 months of intervention. No statistically significant differences between both groups were found for age, height and bone mass. Weight and BMI was lower in the exercise group compared to the control group, increasing lean mass and reducing fat mass. 58.68% of children in the exercise group showed high adherence to the Mediterranean Diet compared to 46.32% of the control group. The exercise group was more concerned about their diet consumed more fiber, vitamin B1, B2, B6, B12, D, Niacin, Folic acid, Fe, Zn, Se and Cu. Triglycerides levels and HDL-cholesterol were higher in the exercise group at the end of the study. Leptin, MCP-1, lipocalin-2, adipsin and PAI-1 levels were lower in the exercise group at the end of the exercise protocol. In contrast, adiponectin and osteocrin markedly increased in the exercise group. Moreover, marked increases were recorded in healthy brain state biomarkers (NGF, BDNF, and irisin) in the exercise group, which could have a positive impact on academic performance. Taken together, all the findings reported are consistent with many benefits of the exercise protocol on adipose tissue and brain molecular function, demonstrating the usefulness of early interventions based on physical activity in children to reduce risk factors related to sedentary lifestyle.
Product Description
Millipore
MILLIPLEX® Human Myokine Magnetic Bead Panel, The Human Myokine Panel, using the Luminex xMAP technology, enables the simultaneous analysis of 15 myokine protein biomarkers in human serum, plasma and cell/tissue culture samples.
Millipore
MILLIPLEX® Human Adipokine Magnetic Bead Panel 1 - Endocrine Multiplex Assay, The Human Adipokine Panel 1, using the Luminex xMAP technology, enables the simultaneous analysis of five biomarkers in human serum or plasma samples.
Millipore
MILLIPLEX® Human Adipokine Magnetic Bead Panel 2 - Endocrine Multiplex Assay, The Human Adipokine Magnetic Bead Panel 2, using the Luminex xMAP technology, enables the simultaneous analysis of nine protein biomarkers in human serum and plasma samples.