Merck
  • Synthesis and structure-activity relationship study of novel cytotoxic carbamate and N-acylheterocyclic bearing derivatives of betulin and betulinic acid.
Synthesis and structure-activity relationship study of novel cytotoxic carbamate and N-acylheterocyclic bearing derivatives of betulin and betulinic acid.
Bioorganic & medicinal chemistry (2010-05-25)
Rita C Santos, Jorge A R Salvador, Silvia Marín, Marta Cascante, João N Moreira, Teresa C P Dinis
ABSTRACT
Chemical transformation studies were conducted on betulin and betulinic acid, common plant-derived lupane-type triterpenes. The concise synthesis, via a stepwise approach, of betulin and betulinic acid carbamate and N-acylheterocyclic containing derivatives is described. All new compounds, as well as betulinic acid were tested in vitro for their cytotoxic activity. Most of the compounds have shown a better cytotoxic profile than betulinic acid, including the synthesized betulin derivatives. Compounds 25 and 32 were the most promising derivatives, being up to 12-fold more potent than betulinic acid against human PC-3 cell lines (IC(50) values of 1.1 and 1.8 microM, respectively).
Sigma-Aldrich
Betulinic acid, technical grade, 90%
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Betulinic acid, ≥98% (HPLC)