Merck
Biodegradable polymeric nanoparticles as drug delivery devices.
Journal of controlled release : official journal of the Controlled Release Society (2001-02-13)
K S Soppimath, T M Aminabhavi, A R Kulkarni, W E Rudzinski
ABSTRACT
This review presents the most outstanding contributions in the field of biodegradable polymeric nanoparticles used as drug delivery systems. Methods of preparation, drug loading and drug release are covered. The most important findings on surface modification methods as well as surface characterization are covered from 1990 through mid-2000.
Sigma-Aldrich
NanoFabTx NanoFlash PEG-PLA drug formulation screening kit, for CIJ synthesis of nanoparticles
Sigma-Aldrich
NanoFabTx PLA-nano, for synthesis of 100 and 200 nm particles
Sigma-Aldrich
NanoFabTx NanoFlash PEG-PCL drug formulation screening kit, for CIJ synthesis of nanoparticles
Sigma-Aldrich
NanoFabTx PEG-PLA drug formulation screening kit, for synthesis of PEGylated nanoparticles
Sigma-Aldrich
NanoFabTx PEGylated nanoparticle formulation screening kit
Sigma-Aldrich
NanoFabTx materials screening kit, for synthesis of polymeric nanoparticles