Merck
  • Oxidative Stress-Induced STIM2 Cysteine Modifications Suppress Store-Operated Calcium Entry.
Oxidative Stress-Induced STIM2 Cysteine Modifications Suppress Store-Operated Calcium Entry.
Cell reports (2020-10-22)
Christine Silvia Gibhardt, Sabrina Cappello, Rajesh Bhardwaj, Romana Schober, Sonja Agnes Kirsch, Zuriñe Bonilla Del Rio, Stefan Gahbauer, Anna Bochicchio, Magdalena Sumanska, Christian Ickes, Ioana Stejerean-Todoran, Miso Mitkovski, Dalia Alansary, Xin Zhang, Aram Revazian, Marc Fahrner, Victoria Lunz, Irene Frischauf, Ting Luo, Daria Ezerina, Joris Messens, Vsevolod Vadimovich Belousov, Markus Hoth, Rainer Arnold Böckmann, Matthias Albrecht Hediger, Rainer Schindl, Ivan Bogeski
ABSTRACT
Store-operated calcium entry (SOCE) through STIM-gated ORAI channels governs vital cellular functions. In this context, SOCE controls cellular redox signaling and is itself regulated by redox modifications. However, the molecular mechanisms underlying this calcium-redox interplay and the functional outcomes are not fully understood. Here, we examine the role of STIM2 in SOCE redox regulation. Redox proteomics identify cysteine 313 as the main redox sensor of STIM2 in vitro and in vivo. Oxidative stress suppresses SOCE and calcium currents in cells overexpressing STIM2 and ORAI1, an effect that is abolished by mutation of cysteine 313. FLIM and FRET microscopy, together with MD simulations, indicate that oxidative modifications of cysteine 313 alter STIM2 activation dynamics and thereby hinder STIM2-mediated gating of ORAI1. In summary, this study establishes STIM2-controlled redox regulation of SOCE as a mechanism that affects several calcium-regulated physiological processes, as well as stress-induced pathologies.
Sigma-Aldrich
Anti-ORAI1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Anti-STIM2 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution