Merck
  • Krebs-cycle-deficient hereditary cancer syndromes are defined by defects in homologous-recombination DNA repair.
Krebs-cycle-deficient hereditary cancer syndromes are defined by defects in homologous-recombination DNA repair.
Nature genetics (2018-07-18)
Parker L Sulkowski, Ranjini K Sundaram, Sebastian Oeck, Christopher D Corso, Yanfeng Liu, Seth Noorbakhsh, Monica Niger, Marta Boeke, Daiki Ueno, Aravind Nambiar Kalathil, Xun Bao, Jing Li, Brian Shuch, Ranjit S Bindra, Peter M Glazer
ABSTRACT
The hereditary cancer syndromes hereditary leiomyomatosis and renal cell cancer (HLRCC) and succinate dehydrogenase-related hereditary paraganglioma and pheochromocytoma (SDH PGL/PCC) are linked to germline loss-of-function mutations in genes encoding the Krebs cycle enzymes fumarate hydratase and succinate dehydrogenase, thus leading to elevated levels of fumarate and succinate, respectively1-3. Here, we report that fumarate and succinate both suppress the homologous recombination (HR) DNA-repair pathway required for the resolution of DNA double-strand breaks (DSBs) and for the maintenance of genomic integrity, thus rendering tumor cells vulnerable to synthetic-lethal targeting with poly(ADP)-ribose polymerase (PARP) inhibitors. These results identify HLRCC and SDH PGL/PCC as familial DNA-repair deficiency syndromes, providing a mechanistic basis to explain their cancer predisposition and suggesting a potentially therapeutic approach for advanced HLRCC and SDH PGL/PCC, both of which are incurable when metastatic.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse