
WHEATON® liquid scintillation vial with attached foil lined urea cap, HDPE

Synonym(s):
WHEATON® liquid scintillation vial with attached foil lined urea cap, glass
Products
Product Category
Capacity (mL)
Diameter (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Showing 1-1 of 1